Learn More
Invitrogen™ SOX21 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 720156
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Mouse and Pig.
The SOX gene family is defined by the Sry-related HMG box domain that mediates sequence-specific DNA binding. A variety of functions are associated with the SOX proteins during development. The highly complex group of Sox genes cluster at least 40 different loci that rapidly diverged in various animal lineages. At present, 30 Sox genes have been identified. Members of this family have been shown to be conserved during evolution and to play key roles during animal development. SOX21 may play a role as an activator of transcription of OPRM1. Overexpression of SOX21 can cause an up-regulation of the OPRM1 distal promoter in mor-expressing neuronal cells.
Specifications
SOX21 | |
Polyclonal | |
Unconjugated | |
Sox21 | |
DNA-binding protein SOX21; HMG-box protein SOX21; Sox21; Sox25; SOX25SOX-A; SOXA; SOX-A; SRY (sex determining region Y)-box 21; SRY box 21; SRY-box 21; SRY-box containing gene 21; transcription factor SOX-21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11166 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
Q9Y651 | |
Sox21 | |
Peptides corresponding to Human SOX21 (aa 80-96, 257-269). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.