Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX3 Polyclonal Antibody

Catalog No. PIPA535983
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535983 Supplier Invitrogen™ Supplier No. PA535983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: MCF-7. Predicted reactivity: Mouse (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX3
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene SOX3
Gene Accession No. P41225, P53784
Gene Alias GHDX; LOC679158; MRGH; PHP; PHPX; similar to SRY (sex determining region Y)-box 3; Sox3; Sox-3; SOXB; SRY (sex determining region Y)-box 3; SRY box 3; SRY-box 3; SRY-box containing gene 3; transcription factor SOX-3
Gene Symbols SOX3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human SOX3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20675, 6658, 679158
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.