Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598169
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598169 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598169 Supplier Invitrogen™ Supplier No. PA598169
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX3 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. Mutations in this gene have been associated with X-linked mental retardation with growth hormone deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX3
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SOX3
Gene Accession No. P41225
Gene Alias GHDX; LOC679158; MRGH; PHP; PHPX; similar to SRY (sex determining region Y)-box 3; Sox3; Sox-3; SOXB; SRY (sex determining region Y)-box 3; SRY box 3; SRY-box 3; SRY-box containing gene 3; transcription factor SOX-3
Gene Symbols SOX3
Host Species Rabbit
Immunogen Recombinant Human Transcription factor SOX-3 protein (4-118AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6658
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.