Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540508 Supplier Invitrogen™ Supplier No. PA540508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PAANNAEISV QLGLEWNKLS EEQKKPYYDE AQKIKEKHRE EFPGWVYQPR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may be involved in the differentiation of developing male germ cells. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX30
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SOX30
Gene Accession No. O94993
Gene Alias LOW QUALITY PROTEIN: transcription factor SOX-30; sex determining region Y-box 30; similar to SRY-box containing gene 30; SOX30; Sox30 protein type II; SRY (sex determining region Y)-box 30; SRY box 30; SRY-box 30; SRY-box containing gene 30; SRY-box transcription factor 30; transcription factor SOX-30
Gene Symbols SOX30
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SOX30.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11063
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.