Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143427 Supplier Invitrogen™ Supplier No. PA5143427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: Zebrafish liver.

SOX32 enables DNA binding activity; DNA-binding transcription factor activity; and protein heterodimerization activity Acts upstream of or within several processes, including circulatory system development; definitive hemopoiesis; and gastrulation Predicted to be located in nucleus Is expressed in several structures, including Kupffer's vesicle; blastoderm; blood island; endoderm; and forerunner cell group [provided by Alliance of Genome Resources, Apr 2022].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX32
Applications Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% Proclin 300; pH 7
Gene Alias 10J3; 226D7; cas; cb527; CG569; SRY (sex determining region Y)-box 32
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of zebrafish Sox32. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.