Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX4 Monoclonal Antibody (CL5634)
GREENER_CHOICE

Catalog No. PIMA531423
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531423 Supplier Invitrogen™ Supplier No. MA531423
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: AGADPSDPLG LYEEEGAG Highest antigen sequence indentity to the following orthologs: Rat - 78%, Mouse - 78%.

SOX genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. SOX genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of SOX genes cluster at a minimum of 40 different loci that rapidly diverged in various animal lineages. At present 30 SOX genes have been identified, and members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone CL5634
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOX4
Gene Accession No. Q06831, Q06945
Gene Alias AA682046; ecotropic viral integration site 16; EVI16; SOX4; Sox-4; SRY (sex determining region Y)-box 4; SRY box 4; SRY-box 4; SRY-box containing gene 4; SRY-box transcription factor 4; SRY-related HMG-box gene 4; transcription factor SOX-4
Gene Symbols SOX4
Host Species Mouse
Immunogen Synthetic peptide corresponding to Human SOX4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20677, 364712, 6659
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.