Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX4 Antibody (SOX4/2540), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279903JF646
Description
SOX4 Monoclonal specifically detects SOX4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SOX4 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SOX4 | |
Recombinant full-length human SOX4 protein (Uniprot: Q06945) | |
0.1 mL | |
Stem Cell Markers, Stem Cells | |
Human | |
IgG2b κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
SOX4/2540 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction