Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX5 Antibody (OTI2A8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274301
Description
SOX5 Monoclonal specifically detects SOX5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SOX5 | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000 | |
L-SOX5, MGC35153, SRY (sex determining region Y)-box 5, transcription factor SOX-5 | |
Mouse | |
83.8 kDa | |
100 μg | |
6660 | |
Human, Mouse, Rat | |
Lyophilized |
Western Blot | |
OTI2A8 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
SOX5 | |
Full length human recombinant protein of human SOX5(NP_008871) produced in HEK293T cell. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction