Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX5 Antibody (OTI2A8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274301H
Description
SOX5 Monoclonal specifically detects SOX5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SOX5 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
SOX5 | |
Full length recombinant protein of human SOX5 (NP_008871) produced in HEK293T cells. | |
0.1 mL | |
6660.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2A8 | |
Western Blot | |
L-SOX5, MGC35153, SRY (sex determining region Y)-box 5, transcription factor SOX-5 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction