Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX5 Monoclonal Antibody (OTI6B5), TrueMAB™, OriGene
SDP

Catalog No. 502540013
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0013 30 μL
50-167-5529 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0013 Supplier OriGene Supplier No. TA504948S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SOX5 Monoclonal antibody specifically detects SOX5 in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen SOX5
Applications Western Blot
Classification Monoclonal
Clone OTI6B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Ftcd
Gene Accession No. O95954, Q91XD4
Gene Alias A730017D01Rik; AI528773; LAMSHF; L-SOX5; L-SOX5B; L-SOX5F; MGC35153; Sox5; Sox-5; Sox5-BLM isoform; Sox5l1; SRY (sex determining region Y)-box 5; SRY box 5; SRY-box 5; SRY-box containing gene 5; SRY-box containing gene 5-like 1; SRY-box transcription factor 5; SRY-related HMG-box 5 protein; transcription factor SOX-5
Host Species Mouse
Immunogen Full length human recombinant protein of human FTCD produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10841, 14317
Target Species Human, Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.