Learn More
SOX5 Mouse anti-Human, Clone: OTI2A8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504932

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.Specifications
SOX5 | |
Monoclonal | |
Unconjugated | |
SOX5 | |
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF | |
Mouse | |
Affinity Chromatography | |
RUO | |
6660 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P35711 | |
SOX5 | |
Full length human recombit protein of human SOX5 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.