Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104343
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104343 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104343 Supplier Invitrogen™ Supplier No. PA5104343
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SOX5.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX5
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SOX5
Gene Accession No. P35710, P35711
Gene Alias A730017D01Rik; AI528773; LAMSHF; L-SOX5; L-SOX5B; L-SOX5F; MGC35153; Sox5; Sox-5; Sox5-BLM isoform; Sox5l1; SRY (sex determining region Y)-box 5; SRY box 5; SRY-box 5; SRY-box containing gene 5; SRY-box containing gene 5-like 1; SRY-box transcription factor 5; SRY-related HMG-box 5 protein; transcription factor SOX-5
Gene Symbols SOX5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SOX5(Accession P35711), corresponding to amino acid residues M90-A140.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140587, 20678, 6660
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.