Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110416
Description
Immunogen sequence: MSSKRPASPY GEADGEVAMV TSRQKVEEEE SDGLPAFHLP LHVSFPNKPH SEEFQPVSLL TQETCGHRTP TSQHNTMEVD GNKVMSSFAP HNSSTSPQKA EEGGRQSGES LSSTALGTPE.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The encoded protein may play a role in chondrogenesis. A pseudogene of this gene is located on chromosome 8. Multiple transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
| SOX5 | |
| Polyclonal | |
| Unconjugated | |
| SOX5 | |
| A730017D01Rik; AI528773; LAMSHF; L-SOX5; L-SOX5B; L-SOX5F; MGC35153; Sox5; Sox-5; Sox5-BLM isoform; Sox5l1; SRY (sex determining region Y)-box 5; SRY box 5; SRY-box 5; SRY-box containing gene 5; SRY-box containing gene 5-like 1; SRY-box transcription factor 5; SRY-related HMG-box 5 protein; transcription factor SOX-5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140587, 20678, 6660 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1.12 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P35710, P35711 | |
| SOX5 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-120 of human SOX5 (NP_6945341). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction