Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX6 Antibody (OTI1E7), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274304R
Description
SOX6 Monoclonal specifically detects SOX6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SOX6 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SOX6 | |
Recombinant protein fragment corresponding to amino acids 1-244 of human SOX6 (NP_201583) produced in E.coli. | |
0.1 mL | |
55553.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI1E7 | |
Western Blot | |
HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction