Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX6 Antibody (OTI1E7), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245917
Description
Ensure accurate, reproducible results in Western Blot
SOX6 Monoclonal specifically detects SOX6 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SOX6 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
Mouse | |
89.6 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
OTI1E7 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
SOX6 | |
Human recombinant protein fragment corresponding to amino acids 1-244 of human SOX6(NP_201583) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
55553 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction