Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534616
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534616 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534616 Supplier Invitrogen™ Supplier No. PA534616
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-34616 targets SOX6 in ChIP, IHC (F, PFA), IHC (P), and WB applications and shows reactivity with Mouse and Rat samples. The PA5-34616 immunogen is synthetic peptide conjugated to KLH derived from within residues 800 to the C-terminus of Mouse SOX6.

The SOX6 gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors, which are known to play vital roles in the regulation of developmental processes and cell fate determination. SOX6, along with its close relatives SOX5 and SOX13, constitutes the SOXD subfamily, which is involved in the regulation of the development of the central nervous system, skeletal system, and cartilage. It functions by binding to specific DNA sequences and modulating the expression of target genes necessary for these developmental processes. SOX6 has been implicated in various functions, such as promoting cartilage formation by acting in conjunction with SOX9 to drive chondrogenesis. In adult neural stem cells, SOX6 is essential for the activation of radial glial-like cells and the generation of new neurons, emphasizing its role in both embryonic development and adult tissue maintenance. The dysregulation of SOX6 has been associated with developmental disorders and its potential involvement in pathologies like cancer, where it influences cell differentiation and proliferation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX6
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene SOX6
Gene Accession No. P40645
Gene Alias AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6
Gene Symbols SOX6
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 800 to the C-terminus of Mouse SOX6.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20679
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.