Learn More
Invitrogen™ SOX6 Polyclonal Antibody
Description
PA1-10882 detects SOX6 in human and mouse samples. This antibody is predicted to recognize rat and chimapanzee SOX6 based on 100% sequence homology. PA1-10882 has been successfully used in Western blot procedures. PA1-10882 immunogen is a synthetic peptide corresponding to residues 780-796 of human and mouse SOX6.
Specifications
Specifications
| Antigen | SOX6 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SOX6 |
| Gene Accession No. | P35712, P40645 |
| Gene Alias | AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6 |
| Gene Symbols | SOX6 |
| Host Species | Rabbit |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.