Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581994
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581994 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581994 Supplier Invitrogen™ Supplier No. PA581994
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TYKPGDNYPV QFIPSTMAAA AASGLSPLQL QQLYAAQLAS MQVSPGAKMP STPQPPNTAG TVSPTGIKNE KRGTSPVTQV KDEAAAQPLN LSSRPKTAEP VKSPTS Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 96%.

The SOX6 gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors, which are known to play vital roles in the regulation of developmental processes and cell fate determination. SOX6, along with its close relatives SOX5 and SOX13, constitutes the SOXD subfamily, which is involved in the regulation of the development of the central nervous system, skeletal system, and cartilage. It functions by binding to specific DNA sequences and modulating the expression of target genes necessary for these developmental processes. SOX6 has been implicated in various functions, such as promoting cartilage formation by acting in conjunction with SOX9 to drive chondrogenesis. In adult neural stem cells, SOX6 is essential for the activation of radial glial-like cells and the generation of new neurons, emphasizing its role in both embryonic development and adult tissue maintenance. The dysregulation of SOX6 has been associated with developmental disorders and its potential involvement in pathologies like cancer, where it influences cell differentiation and proliferation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SOX6
Gene Accession No. P35712
Gene Alias AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6
Gene Symbols SOX6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SOX6. Recombinant protein control fragment (Product #RP-102022).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55553
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.