Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX6 Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993668
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3668 10 μL
50-199-3667 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3668 Supplier Active Motif Supplier No. 61750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Transcriptional activator. Binds specifically to the DNA sequence 5'-AACAAT-3'. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.

Specifications

Antigen SOX6
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH
Gene SOX6
Gene Accession No. P35712
Gene Alias AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; Transcription factor SOX-6
Gene Symbols SOX6
Host Species Rabbit
Immunogen This antibody was raised against a recombinant protein corresponding amino acids 1-20 of human SOX6.
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55553
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.