Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX7 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326090PCP
Description
SOX7 Polyclonal antibody specifically detects SOX7 in Mouse samples. It is validated for Western BlotSpecifications
SOX7 | |
Polyclonal | |
Western Blot | |
MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse SOX7 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
83595 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction