Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SOX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566194
Description
Immunogen sequence: PRRRKSAKAGH SDSDSGAELG PHPGGGAVYK AEAGLGDGHH HGDHTGQTH Highest antigen sequence identity to the following orthologs - mouse 73%, rat 70%.
Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at a minimum of 40 different loci that rapidly diverged in various animal lineages. At present 30 Sox genes have been identified, and members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
Specifications
| SOX8 | |
| Polyclonal | |
| Unconjugated | |
| SOX8 | |
| SOX8; Sox-8; SRY (sex determining region Y)-box 8; SRY box 8; SRY-box 8; SRY-box containing gene 8; transcription factor SOX-8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 30812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P57073 | |
| SOX8 | |
| Recombinant Human SOX8. Recombinant protein control fragment (Product #RP-106880). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction