Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SOX8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598170
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598170 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598170 Supplier Invitrogen™ Supplier No. PA598170
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SOX8 Polyclonal Antibody for Western Blot, ELISA

Sox genes comprise a family of genes that are related to the mammalian sex determining gene SRY. These genes similarly contain sequences that encode for the HMG-box domain, which is responsible for the sequence-specific DNA-binding activity. Sox genes encode putative transcriptional regulators implicated in the decision of cell fates during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at a minimum of 40 different loci that rapidly diverged in various animal lineages. At present 30 Sox genes have been identified, and members of this family have been shown to be conserved during evolution and to play key roles during animal development. Some are involved in human diseases, including sex reversal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.915 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SOX8
Gene Accession No. P57073
Gene Alias SOX8; Sox-8; SRY (sex determining region Y)-box 8; SRY box 8; SRY-box 8; SRY-box containing gene 8; transcription factor SOX-8
Gene Symbols SOX8
Host Species Rabbit
Immunogen Recombinant Human Transcription factor SOX-8 protein (292-386AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30812
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.