Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SOX9 Antibody (SOX9/2287R), DyLight 405, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308592V
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for ELISASpecifications
SOX9 | |
Monoclonal | |
DyLight 405 | |
campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
Recombinant human full-length SOX9 protein (Uniprot: P48436) | |
100 μg | |
Cancer, Stem Cell Markers | |
6662 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
SOX9/2287R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction