Learn More
Invitrogen™ Soybean FTL3 Polyclonal Antibody

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | Soybean FTL3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 4.28 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | FTL3 |
| Gene Accession No. | E3NYP3 |
| Gene Alias | flowering locus T; flowering locus T-like protein 3; FT; FTL3; GmFT2a; LOC100814951; protein HEADING DATE 3A-like |
| Gene Symbols | FTL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.