Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980781
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0781 Supplier Bioss Supplier No. BS10265R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Surfactant protein A is an innate immune system collectin. The gene encoding SP-A map to mouse chromosome 14. Surfactant protein-A, which plays a role in innate host defense in the lung, is also expressed in the Eustachian tube. We report that the frequency of specific surfactant protein-A haplotypes and genotypes differs between children with recurrent otitis media compared with a control population.

Specifications

Antigen SP-A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SFTPA1
Gene Accession No. Q8IWL2
Gene Alias 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC4; Collectin 4; Collectin-4; FLJ50593; FLJ51913; FLJ61144; FLJ77898; FLJ79095; FLJ99559; MGC133365; MGC198590; OTTHUMP00000019928; OTTHUMP00000019929; OTTHUMP00000019930; OTTHUMP00000019931; PSAP; PSP A; PSPA; PSP-A; Pulmonary surfactant-associated protein A; Pulmonary surfactant-associated protein A1; SFTP1; Sftp-1; Sftpa; Sftpa1; SFTPA1B; Sftpl; SP A; SP A1; SPA; SP-A; SPA1; SP-A1; SP-A1 beta; SP-A1 delta; SP-A1 epsilon; SP-A1 gamma; surfactant associated protein A; surfactant associated protein A1; surfactant protein A1; surfactant protein A1 variant AB′D′ 6A; surfactant protein A1 variant AB′D′ 6A2; surfactant protein A1 variant AB′D′ 6A3; surfactant protein A1 variant AB′D′ 6A4; surfactant protein A1 variant ACD′ 6A; surfactant protein A1 variant ACD′ 6A2; surfactant protein A1 variant ACD′ 6A3; surfactant protein A1 variant ACD′ 6A4; surfactant protein A1 variant AD′ 6A; surfactant protein A1 variant AD′ 6A2; surfactant protein A1 variant AD′ 6A3; surfactant protein A1 variant AD′ 6A4; surfactant protein A1B; surfactant pulmonary associated protein A1; surfactant, pulmonary-associated protein A1; surfactant, pulmonary-associated protein A1A; surfactant, pulmonary-associated protein A1B; Surfactant-associated protein 1 (pulmonary surfactant protein SP-A); Surfactant-associated protein 1 (pulmonary surfactant protein, SP-A)
Gene Symbols SFTPA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse SP-A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653509
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.