Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP-B/Surfactant Protein B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP311269
Description
SP-B/Surfactant Protein B Polyclonal specifically detects SP-B/Surfactant Protein B in Human, Porcine samples. It is validated for Western Blot, Immunoassay.Specifications
SP-B/Surfactant Protein B | |
Polyclonal | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Rabbit | |
Protein A purified | |
RUO | |
6439 | |
Store at 4°C. Do not freeze. |
Western Blot, Immunoassay | |
0.1 mg/ml | |
Western Blot, Immunoassay | |
18 kDa pulmonary-surfactant protein, 6 kDa protein, PSP-B, pulmonary surfactant-associated protein B, Pulmonary surfactant-associated proteolipid SPL(Phe), SFTB3, SFTP3, SMDP1, SP-B surfactant, pulmonary-associated protein B, surfactant protein B | |
The polyclonal antibody is raised against porcine SP-B from porcine lungs which has been reconstituted in micelles of lysophosphatidylcholine. | |
50 μg | |
Primary | |
Human, Pig | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction