Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP-D Antibody (1.7) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018518 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018518 0.025 mg
NBP141050 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018518 Supplier Novus Biologicals Supplier No. NBP1410500.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

SP-D Monoclonal specifically detects SP-D in Mouse, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen SP-D
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1.7
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1 ug/ml
Formulation Phosphate Buffered Saline with 0.09% Sodium Azide
Gene Accession No. Q9N1X4
Gene Alias COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4
Gene Symbols SFTPD
Host Species Mouse
Immunogen Purified porcine surfactant protein D
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6441
Test Specificity This antibody detects porcine surfactant protein D (pSP-D), a 138kD member of the collectin family of proteins. Surfactant protein D is thought to play a significant role in the innate immune response against microorganisms within the lungs and at other mucosal surfaces. SP-D is a multifunctional protein which binds selectively to saccharides and lipids on microbial surfaces promoting antimicrobial responses and regulating surfactant lipid homeostasis. SP-D expression is induced in lung surfactant in response to aerogenous bacterial infection, and upon binding to bacteria, promotes phagocytosis and antigen presentation by dendritic cells. Recent studies suggest that pSP-D could serve as a serum biomarker specific for bronchopneumonia in pigs.
Target Species Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.