Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP-D Antibody (111), Alexa Fluor™ 350

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290940AF350
Description
SP-D Monoclonal specifically detects SP-D in Rhesus Macaque samples. It is validated for ELISA.Specifications
SP-D | |
Monoclonal | |
Alexa Fluor 350 | |
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Rhesus SP-D (NP_001035283.1; Met1-Phe375). | |
0.1 mL | |
Cell Cycle and Replication | |
6441 | |
Macaque | |
Purified |
ELISA | |
111 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human SFTPD | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction