Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP-D Antibody (119), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290480AF750
Description
SP-D Monoclonal specifically detects SP-D in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SP-D | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 119 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SP-D (NP_033186.1; Met1-Phe374). | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 6441 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction