Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP-D Antibody (IIIH3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311188
Description
SP-D Monoclonal specifically detects SP-D in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Immunoassay.Specifications
SP-D | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunohistochemistry-Paraffin, Immunoassay | |
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
The monoclonal antibody IIIH3 recognizes the rat surfactant protein D (SP-D). | |
50 μg | |
Cell Cycle and Replication | |
6441 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunoassay | |
IIIH3 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction