Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP-D Rabbit anti-Macaque, Clone: 111, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | SP-D |
---|---|
Clone | 111 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
SP-D Monoclonal specifically detects SP-D in Rhesus Macaque samples. It is validated for ELISA.Specifications
SP-D | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Macaque | |
COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4pulmonary surfactant-associated protein D, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
Recombinant Rhesus SP-D protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
111 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
0.2 um filtered solution in PBS | |
6441 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title