Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Bel-Art™ Biohazard Disposal Bags, 1.5 mil Thick
    
    
    
        
            Shop All SP Bel-Art & SP Wilmad-LabGlass  Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Dimensions (L x W):
                            
                        
                        
                            
                                19 x 14 in. (48 x 36 cm)
                            
                        
                            
                                23 x 19 in. (58 x 48 cm)
                            
                        
                            
                                35 x 25 in. (89 x 64 cm)
                            
                        
                            
                                38 x 31 in. (97 x 79 cm)
                            
                        
                            
                                48 x 38 in. (122 x 97 cm)
                            
                        
                    
                            
                                Capacity (English):
                            
                        
                        
                            
                                13 to 20 gal.
                            
                        
                            
                                2 to 4 gal.
                            
                        
                            
                                20 to 30 gal.
                            
                        
                            
                                40 to 55 gal.
                            
                        
                            
                                5 to 9 gal.
                            
                        
                    Description
- Bright red polypropylene bags include black biohazard symbol
 - Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
 - Sterilization patch darkens when subjected to steam sterilization
 - High temperature steam autoclavable to 135°C (275°F)
 
Specifications
Specifications
| Autoclavable | Autoclavable | 
| Capacity (English) | 20 to 30 gal. | 
| Capacity (Metric) | 95 L | 
| Certifications/Compliance | ASTM 1922 Tear Resistance, ASTM 1709 Dart Impact | 
| Closure Type | Zip Tie | 
| Color | Red | 
| Disposable | Single-use | 
| For Use With (Application) | Used for biohazard waste | 
| Includes | Pre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish | 
| Material | Polypropylene | 
| Show More | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction