Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Clavies™ Autoclave Safe Bag Clips
SCI_ED

Catalog No. s14318 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Color:
Orange

Autoclave safe bag clips can be opened and closed with one hand to easily and securely close the gathered top of an autoclave bag after it has been filled with wastes.

  • Stiff plastic design holds the bag neck closed while allowing release of internal pressure that can develop during the autoclave cycle
  • After autoclaving, the clip is left on the bag for final disposal
  • Suitable for bags ranging in size from 12 x 24 in. to 19 x 23 in.; Use with larger bags is not advised
  • Closed internal dimension: 7/16 in.

Specifications

Autoclavable Autoclavable
Quantity 100
Type Clavies Clip
Material Polypropylene
For Use With (Equipment) Benchtop sized bag or small Scienceware autoclave bag
Color Orange
Jaw Type 2-Prong, 3-Prong
Diameter (Metric) Inside 11 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.