Learn More
Bel Art™ Frigimat™ Junior Dry Ice Maker
Catalog No. 1167525 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Product Type:
Junior Dry Ice Maker
Catalog No. Product Type
11-675-25 Junior Dry Ice Maker
1 options
Catalog No. 11-675-25 Supplier Bel Art™ Supplier No. F388860000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote
Includes
Brass nozzle with polypropylene cone, clamp, heavy-duty cloth bag
Produces dry ice in flake or snow form
  • Screw nozzle directly to cylinder of compressed LCO2; cylinder must have a dip tube
  • When the cylinder valve is opened about 0.75 turn, LCO2 expands into the bag to create dry ice snow; The valve is then closed and the bag is ready to be removed and emptied
  • Secure bag to cone on the nozzle with the clamp
  • After emptying, the bag can be immediately reattached for next filling
  • Makes .77 kg (1.7 lb) of flakes per bag
Order Info
Cone, 5L x 2.312 in. dia. (127 x 59mm) at widest end. Bag, 6 dia. x 12 in. L (15 x 31cm).
Specifications
Type Ice Maker
For Use With (Equipment) Standard CO2 Cylinder
Product Type Junior Dry Ice Maker
Material Brass, Polypropylene
Includes Brass nozzle with Polypropylene cone, clamp, heavy-duty cloth bag
For Use With (Application) Making Dry Ice
Capacity (Metric) 0.77kg