Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ Lab Fridge™ Tray Racks and Cabinet
Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Product Type:
Tray Rack
Tray Rack Cabinet
For Use With (Equipment):
Lab Fridge™ tray cabinet
Lab Fridge™ tray rack
Total Positions:
8
9
Description
- These tray racks eliminate clutter and allow easy stock rotation
- Vacuum-formed white styrene
- Cabinet and Racks sold separately
Lab Fridge Tray Racks
- Suitable for storage of small bottles of blood-typing sera, chemistry controls, enzyme reagents, coagulant reagents, etc.
- Vacuum-formed white styrene is lightweight and easy to clean
- Select 9-channel (Mfr. No. 18661-0000) or 8-channel (18662-0000) tray racks
- 9-channel tray measures 13 x 13 x 1.75 in., with channel width of 1 in.
- 8-channel tray measures 13 x 13 x 1.75 in., with channel width if 1.25 in.
Lab Fridge Tray Rack Cabinet (Mfr. No. 18663-0000)
- Open front cabinet holds up to three Lab Fridge Tray Racks; maximum container height 3 3/4 in. (8.3cm)
- Tray racks slide on side rails and can be easily removed for full access
- Cabinets made of rugged polypropylene that is easy to clean and chemical resistant
Specifications
Specifications
Product Type | Tray Rack Cabinet |
Color | White |
For Use With (Equipment) | Lab Fridge™ tray rack |
Height (English) | 15 in. |
Width (English) | 13 in. |
Width (Metric) | 33.02 cm |
For Use With (Application) | Space saving storage trays and cabinet eliminate clutter and facilitate stock rotation |
Height (Metric) | 38.1 cm |
Dimensions (L x W x H) | 13 x 13 x 15 in. (33.02 x 33.02 x 38.1 cm) |
Length (Metric) | 33.02 cm |
Show More |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction