Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
SCI_ED

Catalog No. p-7078615 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Click to view available options
Dimensions (L x W):
24 x 20 in. (610 x 508 mm)
50 ft. x 20 in.
Color:
Orange
White
Quantity:
100 Sheets/Pk., 6/Pk./Cs.
50 ft./Roll

Catalog No. S42929

Supplier: Bel-Art™ F246750020/EMD

Only null left
Add to Cart
Add to Cart

Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.

  • Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
  • Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
  • Cut to size easily with scissors
  • Roll up and discard in proper waste container after contamination
  • Select orange to identify use with biohazard materials

Specifications

Product Type Bench Protector
Material Polyethylene
Absorbancy 420 mL/m2
Length (English) 24 in.
Length (Metric) 610 mm
Width (English) 20 in.
Width (Metric) 508 mm
Dimensions (L x W) 24 x 20 in. (610 x 508 mm)
Color White
Disposable Single-use
For Use With (Application) Simplify cleanup and safeguard work surfaces from stains, spills and wear
Quantity 100 Sheets/Pk., 6/Pk./Cs.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.