Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ OdoClave™ Deodorant Autoclave Pads

Click to view available options
Scent:
Cherry
Pine
Triple-strength Clove
Color:
Green
Red
Yellow
Description
- Heat-release deodorant works automatically at autoclave temperatures
- Open pouch and place in bag prior to autoclaving
- Three-year shelf life or longer if stored in a sealed container
- Triple strength pads release three times the deodorant of standard pads while costing less than two standard pads
Specifications
Specifications
Product Type | Autoclave Capsule |
Color-coded | No |
Form | Liquid |
Scent | Cherry |
Sterility | Non-sterile |
Autoclavable | Autoclavable |
Color | Red |
Format | Pad |
For Use With (Application) | Autoclave |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction