Learn More
Bel Art™ Space Saver Vacuum Desiccator
SCI_ED
Degas liquids quickly and easily in the laboratory or classroom with these small plastic vacuum desiccators.
$125.00 - $642.00
Specifications
Product Type Vacuum Desiccator
View More Specs
Products 6
Catalog Number Mfr. No. Color HeightEnglish dimHeightMetric Material Price Quantity & Availability  
Catalog Number Mfr. No. Color HeightEnglish dimHeightMetric Material Price Quantity & Availability  
S43283
SCI_ED
View Documents
Bel Art™
F420100000/EMD
Polypropylene, Polycarbonate
Each for $125.00
Case of 4 EA for $490.00
Only null left
Edge
Add to Cart
S43285
SCI_ED
View Documents
Bel Art™
F420200000/EMD
Polypropylene, Polycarbonate
Case of 4 for $642.00
Only null left
Add to Cart
S43284
SCI_ED
View Documents
Bel Art™
F420120000/EMD
Each for $147.00
Only null left
Add to Cart
S43286
SCI_ED
View Documents
Bel Art™
F420220000/EMD
Each for $169.00
Only null left
Add to Cart
S43287
SCI_ED
View Documents
Bel Art™
F420250000/EMD
Polypropylene, Polycarbonate
Each for $223.00
Only null left
Add to Cart
S43288
SCI_ED
View Documents
Bel Art™
F420270000/EMD
Each for $228.00
Only null left
Add to Cart
Description
Description
  • Strong, shatterproof units provide dry storage and protection for sensitive products
  • Lighter and less costly than similarly sized glass models; do not chip like glass
  • Hold a full vacuum (29.9 in. Hg) at room temperature for 24 hours
  • Tapered polypropylene stopcock with PTFE plug accepts tubing with 0.25 in. to 0.375 in. I.D. and allows a controlled entrance of air, preventing unnecessary turbulence
  • Neoprene O-ring assures a tight seal down to 0°C without the need for grease (ring will not be harmed if grease is used)
  • Flat Dome clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
  • All models include a 0.125 in. (0.32cm) thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
  • Clear polycarbonate bottom models offer increased viewing visibility from more angles
  • Models with white polypropylene bottoms can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates (available separately, F42038 series)
  • Models with clear polycarbonate bottoms offer increased visibility from more angles
Certifications
Upper and lower sections, neoprene O-ring, stopcock, polypropylene sample plate
Specifications
Specifications
Vacuum Desiccator
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.