Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Stainless Steel Reagent Digger
SCI_ED

Catalog No. s42990 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Length (Metric):
22.9 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S42990 22.9 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S42990 Supplier Bel-Art™ Supplier No. F367130000/EMD
Only null left
Add to Cart
Add to Cart

Ergonomically designed, time-tested tool, based on confirmed geometry

  • Dislodge stubborn, caked, or crystallized reagents from deep down inside a bottle or vessel that has a minimum neck diameter of 24 mm.
  • Shape facilitates scooping, shoveling, digging and measuring of all powdered, flaked, crystalline or granular media.

Specifications

Product Type Reagent Digger
Material Stainless Steel
Autoclavable Not Autoclavable
For Use With (Application) Scooping, shoveling, digging and measuring of all powdered, flaked, crystalline or granular media from deep down inside a bottle or vessel that has a minimum neck diameter of 24mm
For Use With (Equipment) 2.4cm dia. bottles
Length (English) 9 in.
Length (Metric) 22.9 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.