Learn More
Description
- Made from high-impact polystyrene, FDA grade and sterilized by gamma irradiation
- Comes with flat bottom shape to allow use as weighing boat
- Available in white color
- Available in individual packs for one time use
- Includes certificate of processing for sterilization
Specifications
Specifications
| Sterility | Sterile |
| Autoclavable | Not Autoclavable |
| Shape | Flat Bottom |
| Capacity (Metric) | 2.5 mL |
| Color | White |
| Certifications/Compliance | FDA |
| Disposable | Single-use |
| Product Type | Spoon |
| For Use With (Application) | Ideal for retrieving small, specific volume samples of powders or liquids, provides durability and stiffness when sampling packed powders |
| Includes | Certificate of processing for sterilization |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.