Learn More
Invitrogen™ SP1 Monoclonal Antibody (3C4C3)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549142
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human Caco-2 whole cell, human Jurkat whole cell. IHC: human colorectal adenocarcinoma tissue, human glioma tissue, human laryngeal squamous cell carcinoma tissue, human lung cancer tissue, human ovarian cancer tissue, human thyroid cancer tissue, human tonsil tissue. ICC/IF: Hela cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.
Specifications
SP1 | |
Monoclonal | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P08047 | |
SP1 | |
E. coli-derived human SP1 recombinant protein (Position: Q384-A603). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3C4C3 | |
Unconjugated | |
SP1 | |
1110003E12Rik; AA450830; AI845540; HGNC:11205; SP1; Sp1 transcription factor; Sp1-1; specificity protein 1; trans-acting transcription factor 1; transcription factor Sp1; TSFP1 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
6667 | |
-20°C | |
Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.