Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sp1 Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$159.90 - $633.45
Specifications
| Antigen | Sp1 |
|---|---|
| Concentration | Lot-Specific |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-2874
![]()
|
Active Motif
39810 |
10 μL |
Each for $159.90
|
|
|||||
50-199-2873
![]()
|
Active Motif
39809 |
100 μL |
Each for $633.45
|
|
|||||
Description
SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.Specifications
| Sp1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P08047 | |
| 6667 | |
| This Sp1 antibody was raised against a peptide containing serine 101 of human Sp1. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| SP1 |
| Lot-Specific | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8 | |
| 1110003E12Rik; AA450830; AI845540; HGNC:11205; Sp1; Sp1 transcription factor; Sp1-1; Specificity protein 1; trans-acting transcription factor 1; Transcription factor Sp1; TSFP1 | |
| SP1 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title