Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA595583

Catalog No. PIPA595583


Add to Cart

Description

Description

Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.

SP1 is a transcription factor that can activate or repress transcription in response to physiological and pathological stimuli. Binds with high affinity to GC-rich motifs and regulates the expression of a large number of genes involved in a variety of processes such as cell growth, apoptosis, differentiation and immune responses. Highly regulated by post-translational modifications (phosphorylations, sumoylation, proteolytic cleavage, glycosylation and acetylation). Binds also the PDGFR-alpha G-box promoter. May have a role in modulating the cellular response to DNA damage. Implicated in chromatin remodeling. Plays a role in the recruitment of SMARCA4/BRG1 on the c-FOS promoter. Plays an essential role in the regulation of FE65 gene expression.
Specifications

Specifications

SP1
Polyclonal
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl, 4MG trehalose, 4MG trehalose and 0.05MG sodium azide, 0.05MG sodium azide
O89090, P08047, Q01714
SP1
E.coli-derived human SP1 recombit protein (Position: Q384-A603).
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SP1
Transcription factor Sp1; SP1; TSFP1
Rabbit
Affinity Chromatography
RUO
20683, 24790, 6667
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit