Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sp100, Mouse, Unlabeled, Clone: 2H1.1,

Catalog No. MABC1037MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABC1037MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1037MI Supplier MilliporeSigma™ Supplier No. MABC1037
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nuclear autoantigen Sp-100 (UniProt P23497; also known as Nuclear dot-associated Sp100 protein, Speckled 100kDa)is encoded by the SP100 gene (Gene ID 6672) in human. The SP100 family genes, including SP100, SP110, SP140 and SP140L, encode highly similar proteins mainly expressed in leukocytes. Interferons (IFNs) and other viral infection-related stimuli induce SP100 family gene expression and mutations of SP100 genes are linked to immune diseases and cancers. SP100 colocalizes with promyelocytic leukemia (PML) protein in nuclear bodies (NBs), also known as nuclear domain 10 (ND10), that function to regulate gene transcription via chromatin-based mechanisms and thereby mediate a variety of cellular functions including senescence, apoptosis, protein degradation, telemere stasbility, and defense against viral infections.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sp100
Applications Western Blot
Classification Monoclonal
Clone 2H1.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SP100
Gene Accession No. P23497
Host Species Mouse
Immunogen GST-tagged recombinant human Sp100 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_003104
Test Specificity Clone 2H1.1 targeted epitope region is present in all seven human Sp100 spliced isoforms reported by UniProt (P23497).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.