Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP100 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553476 Supplier Invitrogen™ Supplier No. PA553476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QARTHHNQAS DIIVISSEDS EGSTDVDEPL EVFISAPRSE PVINNDNPLE SNDEKEGQEA TCSRPQIVPE PMDFRKLSTF RESFKKRVIG QDHDFSESSE Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 26%.

This gene encodes a subnuclear organelle and major component of the PML (promyelocytic leukemia)-SP100 nuclear bodies. PML and SP100 are covalently modified by the SUMO-1 modifier, which is considered crucial to nuclear body interactions. The encoded protein binds heterochromatin proteins and is thought to play a role in tumorigenesis, immunity, and gene regulation. Alternatively spliced variants have been identified for this gene; one of which encodes a high-mobility group protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP100
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SP100
Gene Accession No. P23497
Gene Alias A430075G10Rik; DKFZp686E07254; FLJ00340; FLJ34579; lysp100b; nuclear antigen Sp100; nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP 100; SP100; SP100 nuclear antigen; SP100-HMG nuclear autoantigen; speckled 100 kDa
Gene Symbols SP100
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SP100. Recombinant protein control fragment (Product #RP-91589).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.