Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SP110 Antibody (OTI4C1), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274306AF532
Description
SP110 Monoclonal specifically detects SP110 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
SP110 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
SP110 | |
Recombinant protein fragment corresponding to amino acids 232-521 of human SP110 (NP_004501) produced in E.coli. | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI4C1 | |
Western Blot, Immunohistochemistry | |
FLJ22835, IFI41, IFI75, interferon-induced protein 41, 30kD, Interferon-induced protein 41/75, interferon-induced protein 75, 52kD, IPR1, phosphoprotein 41, phosphoprotein 75, SP110 nuclear body protein, Speckled 110 kDa, Transcriptional coactivator Sp110, VODI | |
Mouse | |
0.1 mL | |
3431 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction