Learn More
Invitrogen™ SP110 Monoclonal Antibody (OTI4C1)

Description
SP110 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | SP110 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI4C1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SP110 |
| Gene Accession No. | Q9HB58 |
| Gene Alias | 5031415C07Rik; 52kDa; 5830484A20Rik; ENSMUSG00000075603; IFI41; Ifi75; interferon-induced protein 41, 30kD; interferon-induced protein 41/75; interferon-induced protein 75, 52kD; intracellular pathogen resistance 1; intracellular pathogen resistance protein 1; Ipr1; phosphoprotein 41; phosphoprotein 75; SP110; Sp110 nuclear body protein; speckled 110 kDa; Transcriptional coactivator Sp110; VODI |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.