Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP110 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596701
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596701 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596701 Supplier Invitrogen™ Supplier No. PA596701
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFTMTRAMEE ALFQHFMHQK LGIAYAIHKP FPFFEGLLDN SIITKRMYME SLEACRNLIP VSRVVHNILT QLERTFNLSL LVTLFSQINL REYPNLVTIY RSFKRVGASY ERQSRDTPIL LEAPTGLAEG SSLHTPLALP PPQPPQPSCS PCAPRVSEPG TSSQQSDEIL SESPSPSDPV LPLPALIQEG RSTSVTNDKL TSKMNAEEDS EEMPSLLTST VQVASDNLIP QIRDKEDPQE MPHSPLGSMP EIRDNSPEPN DPEEPQEVSS TPSDKKGKKR KRCIWSTPKR; Positive Samples: NCI-H460, Jurkat, HeLa, Mouse heart, Mouse thymus, Mouse ovary, Mouse liver, Rat brain; Cellular Location: Nucleus.

Susceptibility to tuberculosis (TB) in mice has recently been attributed to the IPR1 gene. IPR1 is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. SP110 is the closest homolog of the IPR1 protein in humans. The IPR1/Sp110 gene product might play a role in integrating signals generated by intracellular pathogens with mechanisms controlling innate immunity, cell death, and pathogenesis. IPR1/Sp110 is up-regulated after infection with M. tuberculosis and required by Anaplasma phagocytophilum for infection of human promyelocytic cells. Defects in Sp110 are a cause of severely impaired resistance to infection by M. tuberculosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP110
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SP110
Gene Accession No. Q3KRF1, Q8BVK9, Q9HB58
Gene Alias 5031415C07Rik; 52kDa; 5830484A20Rik; ENSMUSG00000075603; IFI41; Ifi75; interferon-induced protein 41, 30kD; interferon-induced protein 41/75; interferon-induced protein 75, 52kD; intracellular pathogen resistance 1; intracellular pathogen resistance protein 1; Ipr1; phosphoprotein 41; phosphoprotein 75; SP110; Sp110 nuclear body protein; speckled 110 kDa; Transcriptional coactivator Sp110; VODI
Gene Symbols SP110
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human SP110 (NP_0045013).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109032, 301570, 3431
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.