Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP140 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564325
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564325 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564325 Supplier Invitrogen™ Supplier No. PA564325
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSLLPGEGEE GSDDCSEMCD GEERQEASSS LARRGSVSSE LENHPMNEEG ESEELASSLL YDNVPGAEQS A Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 33%.

Component of the nuclear body, also known as nuclear domain 10, PML oncogenic domain, and KR body. May be involved in the pathogenesis of acute promyelocytic leukemia and viral infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP140
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SP140
Gene Accession No. Q13342
Gene Alias lymphoid-restricted homolog of Sp100; lymphoid-specific SP100 homolog; LYSP100; LYSP100-A; LYSP100-B; nuclear autoantigen Sp-140; Nuclear body protein SP140; SP140; SP140 nuclear body protein; SP140 nuclear body protein family member; speckled 140 kDa
Gene Symbols SP140
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SP140. Recombinant protein control fragment (Product #RP-104218).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11262
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.