Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SP140L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541203
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541203 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541203 Supplier Invitrogen™ Supplier No. PA541203
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: TVDFQAPLLP VTCGGVKGIL HKEKLEQGTL AKCIQTEDGK WFTPMEFEIK Sequence homology: Horse: 80%; Human: 100%; Rat: 79%.

SP140L (SP140 Nuclear Body Protein Like) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include chromatin binding. An important paralog of this gene is SP140. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SP140L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SP140L
Gene Accession No. Q9H930
Gene Alias Nuclear body protein SP140-like protein; SP140 nuclear body protein like; SP140 nuclear body protein-like; SP140L; SP140L-1 protein; SP140L-2 protein; SP140L-3 protein
Gene Symbols SP140L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human LOC93349.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.